Läs mer
Lactoferrin, TRC
Produktkod. 30138185
Change view
Kvantitet:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138185 25 mg
30007270 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30138185 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL11378025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 146897-68-9
Molekylformel C141 H224 N46 O29 S3
Molekylvikt (g/mol) 3123.8
Synonym Lactoferrin, Lactoferricin, Lactoferricin B, MONL 0
IUPAC-namn (2S)-2-[[(2S)-2-[[(2S)-2-[[(2S)-2-[[(2S)-2-[[(3S,9S,12S,15S,18S,21S,24S,27S,30S,33S,36S,39R,44R,47S,50S,53S,56S)-39-[[(2S)-6-amino-2-[[(2S)-2-amino-3-phenyl-propanoyl]amino]hexanoyl]amino]-12,15-bis(4-aminobutyl)-27-(3-amino-3-oxo-propyl)-21,33,36-tris(3-guanidinopropyl)-47-[(1R)-1-hydroxyethyl]-53-(hydroxymethyl)-24,30-bis(1H-indol-3-ylmethyl)-9-isobutyl-3-methyl-50-[(1S)-1-methylpropyl]-18-(2-methylsulfanylethyl)-2,5,8,11,14,17,20,23,26,29,32,35,38,46,49,52,55-heptadecaoxo-41,42-dithia-1,4,7,10,13,16,19,22,25,28,31,34,37,45,48,51,54-heptadecazabicyclo[54.3.0]nonapentacontane-44-carbonyl]amino]-3-methyl-butanoyl]amino]-5-guanidino-pentanoyl]amino]-5-guanidino-pentanoyl]amino]propanoyl]amino]-3-phenyl-propanoic acid
LEDER CC[C@H](C)[C@@H]1NC(=O)[C@H](CO)NC(=O)[C@@H]2CCCN2C(=O)[C@H](C)NC(=O)CNC(=O)[C@H](CC(C)C)NC(=O)[C@H](CCCCN)NC(=O)[C@H](CCCCN)NC(=O)[C@H](CCSC)NC(=O)[C@H](CCCNC(=N)N)NC(=O)[C@H](Cc3c[nH]c4ccccc34)NC(=O)[C@H](CCC(=O)N)NC(=O)[C@H](Cc5c[nH]c6ccccc56)NC(=O)[C@H](CCCNC(=N)N)NC(=O)[C@H](CCCNC(=N)N)NC(=O)[C@H](CSSC[C@H](NC(=O)[C@@H](NC1=O)[C@@H](C)O)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CCCNC(=N)N)C(=O)N[C@@H](CCCNC(=N)N)C(=O)N[C@@H](C)C(=O)N[C@@H](Cc7ccccc7)C(=O)O)NC(=O)[C@H](CCCCN)NC(=O)[C@@H](N)Cc8ccccc8
Specifikationer
Kemiskt namn eller material Lactoferrin
Kvantitet 25 mg
InChI formel InChI=1S/C141H224N46O29S3/c1-11-77(6)111-133(212)186-112(80(9)189)134(213)183-106(130(209)184-110(76(4)5)132(211)176-97(49-32-61-161-141(155)156)120(199)170-93(45-28-57-157-137(147)148)116(195)166-78(7)113(192)180-103(136(215)216)66-82-36-16-13-17-37-82)74-219-218-73-105(182-123(202)90(42-22-25-54-142)167-114(193)87(145)65-81-34-14-12-15-35-81)129(208)173-95(47-30-59-159-139(151)152)118(197)171-96(48-31-60-160-140(153)154)122(201)178-102(68-84-70-163-89-41-21-19-39-86(84)89)127(206)174-98(51-52-108(146)190)124(203)179-101(67-83-69-162-88-40-20-18-38-85(83)88)126(205)172-94(46-29-58-158-138(149)150)119(198)175-99(53-63-217-10)125(204)169-91(43-23-26-55-143)117(196)168-92(44-24-27-56-144)121(200)177-100(64-75(2)3)115(194)164-71-109(191)165-79(8)135(214)187-62-33-50-107(187)131(210)181-104(72-188)128(207)185-111/h12-21,34-41,69-70,75-80,87,90-107,110-112,162-163,188-189H,11,22-33,42-68,71-74,142-145H2,1-10H3,(H2,146,190)(H,164,194)(H,165,191)(H,166,195)(H,167,193)(H,168,196)(H,169,204)(H,170,199)(H,171,197)(H,172,205)(H,173,208)(H,174,206)(H,175,198)(H,176,211)(H,177,200)(H,178,201)(H,179,203)(H,180,192)(H,181,210)(H,182,202)(H,183,213)(H,184,209)(H,185,207)(H,186,212)(H,215,216)(H4,147,148,157)(H4,149,150,158)(H4,151,152,159)(H4,153,154,160)(H4,155,156,161)/t77-,78-,79-,80+,87-,90-,91-,92-,93-,94-,95-,96-,97-,98-,99-,100-,101-,102-,103-,104-,105-,106-,107-,110-,111-,112-/m0/s1
Formel vikt 3121.6629