Läs mer
Lactosamine Hydrochloride, TRC
Produktkod. 30476498
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30476498 5 mg
1 options
Produktkod. 30476498 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL1139305MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Lactosamine Hydrochloride
CAS 203317-42-4
Molekylformel C12H24ClNO10
Kvantitet 5 mg
Synonym 2-Amino-2-deoxylactose Hydrochloride, (2R,3R,4S,5R)-2-Amino-3,5,6-trihydroxy-4-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal Hydrochloride, 2-Amino-2-deoxy-4-O-beta-D-galactopyranosyl-D-glucose Hydrochloride
InChI formel InChI=1S/C12H23NO10.ClH/c13-4(1-14)7(18)11(5(17)2-15)23-12-10(21)9(20)8(19)6(3-16)22-12;/h1,4-12,15-21H,2-3,13H2;1H/t4-,5+,6+,7+,8-,9-,10+,11+,12-;/m0./s1
LEDER Cl.N[C@@H](C=O)[C@@H](O)[C@H](O[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O)[C@H](O)CO
IUPAC-namn (2R,3R,4S,5R)-2-amino-3,5,6-trihydroxy-4-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhexanal;hydrochloride
Molekylvikt (g/mol) 377.77
Formel vikt 377.1089