Läs mer
Lactosazone, TRC
Produktkod. 30468372
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30468372 100 mg
1 options
Produktkod. 30468372 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL113925100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Lactosazone
CAS 4746-17-2
Molekylformel C24H32N4O9
Kvantitet 100 mg
Synonym 4-O-beta-D-Galactopyranosyl-D-arabino-hexosulose Bis(phenylhydrazone), Lactose Phenylosazone, NSC 56106, 4-O-beta-D-Galactopyranosyl-D-arabino-Hexos-2-ulose Bis(phenylhydrazone)
InChI formel InChI=1S/C24H32N4O9/c29-12-17(31)23(37-24-22(35)21(34)20(33)18(13-30)36-24)19(32)16(28-27-15-9-5-2-6-10-15)11-25-26-14-7-3-1-4-8-14/h1-11,17-24,26-27,29-35H,12-13H2/b25-11+,28-16+
LEDER OCC(O)C(O[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O)C(O)\C(=N\Nc2ccccc2)\C=N\Nc3ccccc3
Molekylvikt (g/mol) 520.53
Formel vikt 520.2169