Learn More
LAIR1 Mouse anti-Human, Unconjugated, Clone: 1A5D1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67220-1-IG-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16839745
  • 3313.85 SEK / 150µL
Estimated Shipment: 03-10-2022
to see stock.
Description
Description
The LAIR1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human LAIR1. This antibody recognizes human antigen. The LAIR1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
LAIR1
Monoclonal
1000 μg/mL
Mouse
Primary
Human
ELISA, Immunofluorescence, Immunohistochemistry, Western Blot
1A5D1
Unconjugated
150 μL
3903
IgG2a