Beskrivning
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifikationer
Specifikationer
| Kemiskt namn eller material | lambda-Cyhalothrin |
| CAS | 91465-08-6 |
| CAS Max % | ≤ 100% |
| Flampunkt | 83°C |
| Färg | Colorless |
| Smältpunkt | 49.2°C |
| Kokpunkt | 270°C |
| Synonym | (R)-cyano(3-phenoxyphenyl)methyl (1S,3S)-rel-3-[(1Z)-2-chloro-3,3,3-trifluoro-1-propen-1-yl]-2,2-dimethylcyclopropanecarboxylate |
| Molekylvikt (g/mol) | 449.9 g/mol |
| Kvantitet | 100 mg |