missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Lambda Light Chain Antibody (ICO-106), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-33308DL594
4998.00 SEK valid until 2025-12-16
BÄSTA PRIS på kampanj! Använd kampanjkod: "24090" för att få ditt kampanjpris.
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Lambda Light Chain Monoclonal specifically detects Lambda Light Chain in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifikationer
| Lambda Light Chain | |
| Monoclonal | |
| DyLight 594 | |
| IGLC, immunoglobulin lambda constant group | |
| Human IgG was used as immunogen to generate the Lambda antibody. | |
| 0.1 mL | |
| Adaptive Immunity, Immunology | |
| 3537 | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| ICO-106 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| This monoclonal antibody is specific to lambda light chain of immunoglobulin and shows no cross-reaction with lambda light chain or any of the five heavy chains. In mammals, the two light chains in an antibody are always identical, with only one type of light chain, kappa or lambda. The ratio of Kappa to Lambda is 70:30. However, with the occurrence of multiple myeloma or other B-cell malignancies this ratio is disturbed. Antibody to the lambda light chain is reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkins lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is malignant. | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering