missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ LAMC1 (Human) Recombinant Protein
Human LAMC1 full-length ORF ( AAH15586, 1 a.a. - 38 a.a.) recombinant protein with GST-tag at N-terminal.
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH15586 |
---|---|
Gene ID (Entrez) | 3915 |
Name | laminin, gamma 1 (formerly LAMB2) |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16127311
|
Abnova™
H00003915-P01.10UG |
10 μg |
4515.00 SEK
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16137311
|
Abnova™
H00003915-P01.25UG |
25 μg |
6845.00 SEK
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Sequence: MNKRRTSHRIWKNKLPEYMRRPKGPVTKLWRSMPAWLS
Specifications
AAH15586 | |
laminin, gamma 1 (formerly LAMB2) | |
125% SDS-PAGE Stained with Coomassie Blue | |
LAMB2, MGC87297 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
3915 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LAMC1 | |
GST |