Learn More
Brady™ Laminated Polyester: ISO Safety Sign - Wear eye protection
Product Code. 15614457 Shop All Brady Products
Change view
Click to view available options
Diameter (Metric):
50 mm
100 mm
200 mm
315 mm
Unit Size:
Each
Pack of 2
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
15614457 50 mm Pack of 2
15356827 100 mm Each
15316388 200 mm Each
15624457 315 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15614457 Supplier Brady™ Supplier No. 818418
  • ONLINE EXCLUSIVE PRICE
    69.33 SEK
    57.71 SEK / Paket med 2
    Save 11.62 SEK
    16% Av
I lager
Sign compliant to ISO 7010.
  • Laminated polyester
  • For indoor and outdoor use
Specifications
Certifications/Compliance ISO 7010
Alternate Part Number PIC M004-Dia 050-PE-SHEET/2
Finish Gloss
Material Polyester
Quantity 2/Pk.
Adhesive Type Permanent Acrylic
Diameter (Metric) 50 mm
For Use With (Application) Indoor and Outdoor
Thickness 0.099 mm
Temperature Range (Metric) -40°C to 110°C
Shape Round
Languages Global
Color White on Blue