Learn More
Laminin-211 Recombinant Polyclonal Antibody (22HCLC), Invitrogen™
Rabbit Recombinant Polyclonal Antibody
Brand: Invitrogen 711233
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with Monkey, Rabbit, Cat and Bovine. Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.
Laminin-211 Recombinant Polyclonal antibody specifically detects Laminin-211 in Human samples. It is validated for Immunocytochemistry.Specifications
Laminin-211 | |
Recombinant Polyclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P24043 | |
LAMA2 | |
Protein corresponding to Human LAMA2 (aa 1170-1380) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry | |
22HCLC | |
Unconjugated | |
LAMA2 | |
5830440B04; dy; dystrophia muscularis; Lama2; laminin alpha-2; laminin M; Laminin M chain; laminin subunit alpha 2; laminin subunit alpha-2; laminin, alpha 2; laminin-12 subunit alpha; Laminin-2 subunit alpha; laminin-4 subunit alpha; LAMM; mer; merosin; Merosin heavy chain; mKIAA4087 | |
Rabbit | |
Protein A | |
RUO | |
3908 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |