Läs mer
Lamivudine-15N2,13C, TRC
Produktkod. 30133370
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133370 10 mg
30008922 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133370 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL17250210MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1217746-03-6
Molekylformel 13C C7 H11 15N2 N O3 S
Molekylvikt (g/mol) 232.24
Synonym Lamivudine-15N2,13C, cis-Lamivudine-15N2,13C, 4-Amino-1-[(2RS,5SR)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl]pyrimidin-2(1H)-one-1,3-15N2,2-13C, [1,3-15N2,2-13C]-4-Amino-1-[(2RS,5SR)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl]pyrimidin-2(1H)-one, cis-2-Hydroxymethyl-5-(cytosin-1-yl-1,3-15N2,2-13C)-1,3-oxathiolane
IUPAC-namn 4-amino-1-[(2R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl](2^{13}C,1,3-^{15}N_{2})pyrimidin-2-one
LEDER NC1=[15N][13C](=O)[15N](C=C1)[C@@H]2CS[C@H](CO)O2
Specifikationer
Kemiskt namn eller material Lamivudine-1,3-15N2,2-13C
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C8H11N3O3S/c9-5-1-2-11(8(13)10-5)6-4-15-7(3-12)14-6/h1-2,6-7,12H,3-4H2,(H2,9,10,13)/t6-,7+/m0/s1/i8+1,10+1,11+1
Formel vikt 232.0495
Procent renhet >95%