Läs mer
Lamivudine Monophosphate Triethylammonium Salt, TRC
Produktkod. 30466017
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30466017 10 mg
1 options
Produktkod. 30466017 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL17252010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Lamivudine Monophosphate Triethylammonium Salt
CAS 2724549-70-4
Molekylformel C8H12N3O6PS:[C6H15N]x
Kvantitet 10 mg
Synonym 4-Amino-1-[(2R,5S)-2-[(phosphonooxy)methyl]-1,3-oxathiolan-5-yl]-2(1H)-pyrimidinone Triethylammonium Salt
InChI formel InChI=1S/C8H12N3O6PS.C6H15N/c9-5-1-2-11(8(12)10-5)6-4-19-7(17-6)3-16-18(13,14)15;1-4-7(5-2)6-3/h1-2,6-7H,3-4H2,(H2,9,10,12)(H2,13,14,15);4-6H2,1-3H3/t6-,7+;/m0./s1
LEDER CCN(CC)CC.NC1=NC(N([C@@H]2CS[C@H](COP(O)(O)=O)O2)C=C1)=O
IUPAC-namn triethylamine ((2R,5S)-5-(4-amino-2-oxopyrimidin-1(2H)-yl)-1,3-oxathiolan-2-yl)methyl phosphate
Molekylvikt (g/mol) 410.43