Läs mer
Lamivudine (R)-Sulfoxide, TRC
Produktkod. 30481637
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30481637 10 mg
30475632 50 mg
30475785 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30481637 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL17247510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 160552-54-5
Molekylformel C8 H11 N3 O4 S
Molekylvikt (g/mol) 245.26
Synonym Lamivudine (R)-Sulfoxide, GI138870X
IUPAC-namn 4-amino-1-[(2R,3R,5S)-2-(hydroxymethyl)-3-oxo-1,3-oxathiolan-5-yl]pyrimidin-2-one
LEDER NC1=NC(=O)N(C=C1)[C@@H]2C[S@@](=O)[C@H](CO)O2
Specifikationer
Kemiskt namn eller material 4-Amino-1-[(2R,3R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl]pyrimidin-2(1H)-one S-Oxide
Kvantitet 10 mg
InChI formel InChI=1S/C8H11N3O4S/c9-5-1-2-11(8(13)10-5)6-4-16(14)7(3-12)15-6/h1-2,6-7,12H,3-4H2,(H2,9,10,13)/t6-,7+,16+/m0/s1
Formel vikt 245.047