Läs mer
Lamotrigine-13C3, TRC
Produktkod. 18010640
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18010640 1 mg
18001640 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18010640 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL1732521MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1188265-38-4
Molekylformel 13C3 C6 H7 Cl2 N5
Molekylvikt (g/mol) 259.07
Synonym 1,2,4-Triazine-3,5-diamine-3,5,6-13C3, 6-(2,3-dichlorophenyl)- (ACI), 6-(2,3-Dichlorophenyl)-1,2,4-triazine-3,5-diamine-3,5,6-13C3 (ACI), Lamotrigine-13C3, Lamotrigine-13C3 (1,2,4-triazine-13C3)
IUPAC-namn 6-(2,3-dichlorophenyl)-(3,5,6-^{13}C_{3})1,2,4-triazine-3,5-diamine
LEDER N[13c]1nn[13c]([13c](N)n1)c2cccc(Cl)c2Cl
Specifikationer
Kemiskt namn eller material Lamotrigine-13C3
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C9H7Cl2N5/c10-5-3-1-2-4(6(5)11)7-8(12)14-9(13)16-15-7/h1-3H,(H4,12,13,14,16)/i7+1,8+1,9+1
Formel vikt 258.0179
Procent renhet >95%