Läs mer
Lamotrigine EP Impurity C, Mikromol™
Produktkod. 30051580
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30051580 25 mg
30049130 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30051580 Leverantör Mikromol Leverantörsnummer MM0922.030025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Lamotrigine EP Impurity C
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2Z)-[2-(Diaminomethylidene)diazanylidene](2,3-dichlorophenyl)acetonitrile
CAS 94213-23-7
Analyt- eller komponentnamn (2Z)-[2-(Diaminomethylidene)diazanylidene](2,3-dichlorophenyl)acetonitrile
Frakt skick Room Temperature
Molekylformel C9 H7 Cl2 N5
Certifikat och efterlevnad ISO 17025
Synonym Lamotrigine Imp. C (EP), (2Z)-[2-(Diaminomethylidene)diazanylidene](2,3-dichlorophenyl)acetonitrile, Lamotrigine Impurity C
InChI formel InChI=1S/C9H7Cl2N5/c10-6-3-1-2-5(8(6)11)7(4-12)15-16-9(13)14/h1-3H,(H4,13,14,16)/b15-7+
LEDER NC(=N\N=C(/C#N)\c1cccc(Cl)c1Cl)N
IUPAC-namn (1Z)-2,3-dichloro-N-(diaminomethylideneamino)benzenecarboximidoyl cyanide
Molekylvikt (g/mol) 256.09
Kvantitet 25 mg
Formel vikt 255.0079 g/mol