missing translation for 'onlineSavingsMsg'
Läs mer

Lamotrigine N2-Oxide, TRC

Produktkod. 30133371
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133371 25 mg
30132903 10 mg
30006506 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30133371 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL17328025MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 136565-76-9
Molekylformel C9 H7 Cl2 N5 O
Molekylvikt (g/mol) 272.09
Synonym 1,2,4-Triazine-3,5-diamine, 6-(2,3-dichlorophenyl)-, 2-oxide, Lamotrigine N2-Oxide
IUPAC-namn 6-(2,3-dichlorophenyl)-2-oxido-1,2,4-triazin-2-ium-3,5-diamine
LEDER Nc1nc(N)[n+]([O-])nc1c2cccc(Cl)c2Cl

Specifikationer

Kemiskt namn eller material Lamotrigine N2-Oxide
Kvantitet 25 mg
InChI formel InChI=1S/C9H7Cl2N5O/c10-5-3-1-2-4(6(5)11)7-8(12)14-9(13)16(17)15-7/h1-3H,(H4,12,13,14)
Formel vikt 271.0028
Produkttitel
Välj ett problem

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.