Läs mer
Lanabecestat, MedChemExpress
Produktkod. 16489696
Encompass_Preferred
Change view
Kvantitet:
2 mg
5 mg
10 mg
50 mg
100 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16489696 50 mg
16484436 100 mg
16460036 2 mg
16453148 10 mg
16405887 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Produktkod. 16489696 Leverantör Medchem Express Leverantörsnummer HY100740
MedChemExpress Lanabecestat (AZD3293) is a potent, orally active and blood-brain barrier penetrating BACE1 inhibitor with a Ki of 0.4 nM. Lanabecestat is used for the research of Alzheimer's disease.
Lanabecestat (AZD3293) is a potent, orally active and blood-brain barrier penetrating BACE1 inhibitor with a Ki of 0.4 nM. Lanabecestat is used for the research of Alzheimer's disease.
Specifikationer
Kemiskt namn eller material Lanabecestat
CAS 1383982-64-6
Färg Earth Yellow
Anteckningar om renhetsgrad Research
Molekylformel C26H28N4O
Kvantitet 50 mg
Synonym AZD3293 LY3314814
Löslighetsinformation DMSO : ≥ 100 mg/mL (242.41 mM)
LEDER CC#CC1=CC(C2=CC([C@@]34N=C(C)C(N)=N4)=C(C[C@@]53CC[C@@H](OC)CC5)C=C2)=CN=C1
Molekylvikt (g/mol) 412.53
Formel vikt 412.53
Procent renhet 98.55%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Neuroscience-Neurodegeneration