Läs mer
Logistik™ Lancer-Style Chemical Splash Laboratory Coats
Produktkod. 12814382
Change view
Storlek:
Small
Medium
Large
Förpackningsstorlek:
Varje
Produktkod. Storlek unitSize
12814382 Large Varje
12834382 Small Varje
12824382 Medium Varje
3 options
Produktkod. 12814382
Brand: Logistik™ 849AE2WH108CM
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Provide the answer to full frontal protection with Logistik™ Lancer-Style Chemical Splash Laboratory Coats The wrap-over style Lancer front coat helps repel liquid chemicals and provides the wearer with an excellent level of protection since the use of chemicals in a laboratory can prove a constant hazard to standard style open neck coats.
  • Full frontal protection
  • Chem-Tech material
  • Fluorochemical liquid repellent
Specifikationer
Produkttyp General Purpose Lab Coat
Storlek Large
Kön Unisex
Material Polyester, Carbon
Färg White
Manschettstil Stud-fastened
Disponibel Reusable
Sterilitet Non-sterile
Stil Lancer
Antal fickor 2
Krage typ Open
Avsluta Fluorochemical Liquid Repellent
Storlek (metrisk) bröst 106.68 to 111.76 cm
Certifikat och efterlevnad CE EN 13034
Längd (metrisk) 108 cm
Konstruktion 65% Polyester, 35% Cotton