Learn More
Lanoconazole, TRC
Product Code. 30468400
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity Unit Size
30468400 100 mg 100mg
1 options
Product Code. 30468400 Supplier Toronto Research Chemicals Supplier No. TRCL174500100MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Lanoconazole
CAS 101530-10-3
Molecular Formula C14 H10 Cl N3 S2
Quantity 100 mg
Synonym 1H-Imidazole-1-acetonitrile, α-[4-(2-chlorophenyl)-1,3-dithiolan-2-ylidene]-, (E)-(±)-, (αE)-α-[4-(2-Chlorophenyl)-1,3-dithiolan-2-ylidene]-1H-imidazole-1-acetonitrile, 1H-Imidazole-1-acetonitrile, α-[4-(2-chlorophenyl)-1,3-dithiolan-2-ylidene]-, (E)-, Astat, Latoconazole, NND 318, TJN 318
InChI Formula InChI=1S/C14H10ClN3S2/c15-11-4-2-1-3-10(11)13-8-19-14(20-13)12(7-16)18-6-5-17-9-18/h1-6,9,13H,8H2/b14-12+
SMILES Clc1ccccc1C2CS\C(=C(\C#N)/n3ccnc3)\S2
IUPAC Name (2E)-2-[4-(2-chlorophenyl)-1,3-dithiolan-2-ylidene]-2-imidazol-1-ylacetonitrile
Molecular Weight (g/mol) 319.83
Formula Weight 319.0005