Läs mer
Lanreotide Acetate, TRC
Produktkod. 30134530
Change view
Kvantitet:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134530 100 mg
30133732 25 mg
30008509 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134530 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL174600100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 127984-74-1
Synonym L-Threoninamide, 3-(2-naphthalenyl)-D-alanyl-L-cysteinyl-L-tyrosyl-D-tryptophyl-L-lysyl-L-valyl-L-cysteinyl-, cyclic (2→7)-disulfide, acetate (salt) (9CI)∣1,2-Dithia-5,8,11,14,17-pentaazacycloeicosane, cyclic peptide deriv.∣BIM 23014C∣Lanreotide acetate∣Somatulina∣Somatuline
Specifikationer
Renhet >95% (HPLC)
Formatera Neat
Molekylformel C56H73N11O12S2
MDL-nummer MFCD00873576
Molekylvikt (g/mol) 1156.38, 1156.39
InChI-nyckel DEXPIBGCLCPUHE-UHFFFAOYNA-N
LEDER CC(O)=O.CC(C)C1NC(=O)C(CCCCN)NC(=O)C(CC2=CNC3=CC=CC=C23)NC(=O)C(CC2=CC=C(O)C=C2)NC(=O)C(CSSCC(NC1=O)C(=O)NC(C(C)O)C(N)=O)NC(=O)C(N)CC1=CC2=CC=CC=C2C=C1
IUPAC-namn 2-({19-[2-amino-3-(naphthalen-2-yl)propanamido]-10-(4-aminobutyl)-16-[(4-hydroxyphenyl)methyl]-13-[(1H-indol-3-yl)methyl]-6,9,12,15,18-pentaoxo-7-(propan-2-yl)-1,2-dithia-5,8,11,14,17-pentaazacycloicosan-4-yl}formamido)-3-hydroxybutanamide; acetic acid