missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LanthaScreen™ Eu-anti-His Tag Antibody Kit
LanthaScreen™ Eu-anti-His Tag Antibody Kit
Brand: Invitrogen PV5597
Additional Details : Weight : 0.00010kg
Description
LanthaScreen™ europium-labeled anti-His-tag antibodies are used to indirectly label proteins tagged with 6XHis. This can be advantageous when working with partially purified protein, or in cases in which direct labeling of the protein using amine-reactive Eu chelates negatively impacts protein activity. These antibodies can be used with our Kinase Tracers to conduct LanthaScreen™ Eu Kinase Binding Assays.Specifications
His | |
0.25 mg/mL, 0.25 mg/ml, 0.25 mg⁄ml | |
150kDa | |
RUO, RUO | |
Primary | |
Europium-labeled anti-His tag antibody in HEPES buffered saline. Store at -20°C. | |
Liquid | |
Microplate Reader | |
His | |
LanthaScreen™ |
Kinase Assay, Kinase Assay | |
Europium | |
1 mg | |
Target and Lead Identification and Validation | |
1 Kit(s) | |
150 kDa | |
340⁄665 | |
Dry Ice | |
TR-FRET |
For Research Use Only. Not for use in diagnostic procedures.