Läs mer
Thermo Scientific™ LanthaScreen™ Tb-anti-Histone H3K4me3 Antibody
Beskrivning
- The LanthaScreen™ Tb-anti-Histone H3K4me3 Antibody is a key component of the BacMam Histone H3K4me3 Cellular Assay (A14163)
- The LanthaScreen™ Histone H3K4me3 antibody allows investigation of trimethylation at lysine 4 on histone H3
- This enables screening for potential inhibitors of methyl transferases and demethylases associated with Lys4 trimethylation in their natural complexes
- For research use only
- Not for human or animal therapeutic or diagnostic use
LanthaScreen™ Tb-anti-Histone H3K4me3 Antibody Lets You:
- Identify more relevant inhibitors since the methyl transferases and demethylases are in their natural protein complexes
- Conserve precious cell samples with the miniaturizable homogenous assay format
- Improve data quality with the advantages of TR-FRET
Specifikationer
Specifikationer
| Koncentration | 3500 nM, 0.5 mg/mL |
| Konjugera | Tb (Terbium)/Lanthanide Chelates |
| Kvantitet | 1 mg |
| Innehåll och lagring | Store the LanthaScreen™ Tb-anti-histone H3K4me3 Antibody at -20°C. |
| Form | Liquid |
| Frakt skick | Dry Ice |
| Måländringstillstånd | Methylated |