missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LanthaScreen™ Tb-Ubiquitin
LanthaScreen™ Tb-Ubiquitin
5160.00 SEK - 20650.00 SEK
Specifications
Conjugate | Terbium (Tb) |
---|---|
Content And Storage | Terbium-labeled ubiquitin in HEPES buffered saline. Store at -20°C. |
Detection Method | Fluorescent |
Excitation/Emission | 340⁄490, 520 |
For Use With (Equipment) | Microplate Reader |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11505016
|
Invitrogen
PV4375 |
5μg |
5160.00 SEK
5µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11515016
|
Invitrogen
PV4376 |
25μg |
20650.00 SEK
25µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Ubiquitin has been labeled with the LanthaScreen™ Terbium chelate for use in high throughput assays to monitor protein ubiquitination. Due to the method used to label the ubiquitin protein, all of the lysines with the protein are available for further conjugation to form poly-ubiquitin chains.For use in combination with FRET acceptors including Fluorescein-ubiquitin (PV4377) or a fluorescein-labeled antibody.
Specifications
Terbium (Tb) | |
Fluorescent | |
Microplate Reader | |
Lanthanide Chelates | |
Dry Ice | |
TR-FRET | |
LanthaScreen™ |
Terbium-labeled ubiquitin in HEPES buffered saline. Store at -20°C. | |
340⁄490, 520 | |
Tb (Terbium) | |
End Point | |
Protein-Based Substrate | |
Liquid |
For Research Use Only. Not for use in diagnostic procedures.