Läs mer
Lapatinib-13C2,15N Ditosylate, TRC
Produktkod. 30466016
Change view
Kvantitet:
0.5 mg
Produktkod. Kvantitet
30466016 0.5 mg
1 options
Produktkod. 30466016 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL1758020.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Lapatinib-13C2,15N Ditosylate
CAS 1246819-08-8
Molekylformel C4113C2H42ClFN315NO10S3
Kvantitet 0.5 mg
Synonym N-[3-Chloro-4-[(3-fluorophenyl)methoxy]phenyl]-6-[5-[[[2-(methylsulfonyl)ethyl]amino]methyl]-2-furanyl]-4-quinazolinamine Bis(4-methylbenzenesulfonate)-13C2,15N Ditosylate, GW-572016F-13C2,15N, Tykerb-13C2,15N Ditosylate
InChI formel InChI=1S/C29H26ClFN4O4S.C7H8O3S/c1-40(36,37)12-11-32-16-23-7-10-27(39-23)20-5-8-26-24(14-20)29(34-18-33-26)35-22-6-9-28(25(30)15-22)38-17-19-3-2-4-21(31)13-19;1-6-2-4-7(5-3-6)11(8,9)10/h2-10,13-15,18,32H,11-12,16-17H2,1H3,(H,33,34,35);2-5H,1H3,(H,8,9,10)/i11+1,12+1,32+1;
LEDER CC1=CC=C(S(=O)(O)=O)C=C1.FC2=CC(COC(C(Cl)=C3)=CC=C3NC4=NC=NC5=CC=C(C6=CC=C(C[15NH][13CH2][13CH2]S(C)(=O)=O)O6)C=C54)=CC=C2
Molekylvikt (g/mol) 928.44