Läs mer
Lappaconitine Hydrobromide, TRC
Produktkod. 30478890
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30478890 250 mg
1 options
Produktkod. 30478890 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL175855250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Lappaconitine Hydrobromide
CAS 97792-45-5
Molekylformel C32H45BrN2O8
Kvantitet 250 mg
Synonym 2H-12,3,6a-Ethanylylidene-7,9-methanonaphth[2,3-b]azocine, aconitane-4,8,9-triol deriv. Hydrobromide, Acetyl-10-deoxysepaconitine Hydrobromide
InChI formel InChI=1S/C32H44N2O8.BrH/c1-6-34-16-29(42-28(36)18-9-7-8-10-21(18)33-17(2)35)12-11-25(40-4)31-23(29)14-20(26(31)34)30(37)15-22(39-3)19-13-24(31)32(30,38)27(19)41-5;/h7-10,19-20,22-27,37-38H,6,11-16H2,1-5H3,(H,33,35);1H/t19-,20+,22+,23-,24+,25+,26-,27+,29-,30+,31+,32+;/m1./s1
LEDER O[C@]1([C@]2(O)[C@@]3([H])CC([C@@H](OC)C1)[C@@H]2OC)C4C[C@@]5([H])C63[C@@]4([H])N(CC)C[C@@]5(OC(C7=CC=CC=C7NC(C)=O)=O)CC[C@H]6OC.Br
Molekylvikt (g/mol) 665.61
Formel vikt 664.2359