Learn More
CleanSpace Technology™ Large Case Particulate Pre-Filters
Suitable for protection against airborne particulates (dust, mists and fumes). Designed to remove course particles and enhance the life of the Particulate P3 Filter on PAF-0037 and Combined Filters. The Pre-Filter should be changed after each use.
Brand: CleanSpace Technology™ PAF-0057
Additional Details : Weight : 0.05200kg
Description
- Used with the revolutionary CleanSpace - a light weight PAPR with no hoses/belts
- Materials: Spun polymer fibers
- Easy and quickly fitted and removed from the power unit
- Sold in packs of 20
- Compatible with all CleanSpace Respirators
Specifications
Prefilter |