missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical™ Larger Fittings Ferrules

Provides leak-free system connections. Trajan Scientific and Medical Larger Fittings Ferrules leads to improved chromatography for non-MS systems.

695.00 SEK

Specifications

Material 100% Graphite
View More Specs

Products 2
Product Code Brand Diameter (Metric) Inside Width (Metric) Length (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Inside Width (Metric) Length (Metric) Price Quantity & Availability  
15278863
View Documents Promotion Details
Trajan Scientific and Medical™
072613
12.7 mm 17.2 mm 6.4 mm
695.00 SEK
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Add to basket
 
15248863
View Documents Promotion Details
Trajan Scientific and Medical™
072605
5 mm 8 mm 4.5 mm
695.00 SEK
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Add to basket
 
Description

Description

  • Easy to use
  • Forms a reliable seal
  • Reusable
  • Soft material, forms a soft grip to the capillary column
  • Low emissions
Specifications

Specifications

100% Graphite
Videos
SDS
Documents

Documents

Product Certifications

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.