Learn More
Trajan Scientific and Medical™ Larger Fittings Ferrules
Provides leak-free system connections. Trajan Scientific and Medical Larger Fittings Ferrules leads to improved chromatography for non-MS systems.
Brand: Trajan Scientific and Medical™ 072605
Additional Details : Weight : 0.01000kg
Description
- Easy to use
- Forms a reliable seal
- Reusable
- Soft material, forms a soft grip to the capillary column
- Low emissions
Specifications
8 mm | |
4.5 mm | |
5 mm |
100% Graphite | |
Larger Ferrule 5 mm Fitting |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.