Läs mer
Larixol, TRC
Produktkod. 30468702
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30468702 500 mg
30483209 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30468702 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL175950500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1438-66-0
Molekylformel C20H34O2
Molekylvikt (g/mol) 306.48
Synonym (alphaS,1S,4S,4aS,8aR)-alpha-Ethenyldecahydro-4-hydroxy-alpha,5,5,8a-tetramethyl-2-methylene-1-naphthalenepropanol, [1S-[1alpha(R*),4beta,4abeta,8aalpha]]-alpha-Ethenyldecahydro-4-hydroxy-alpha,5,5,8a-tetramethyl-2-methylene-1-naphthalenepropanol, (+)-Larixol
IUPAC-namn (1S,4S,4aR,8aS)-4-[(3S)-3-hydroxy-3-methylpent-4-enyl]-4a,8,8-trimethyl-3-methylidene-2,4,5,6,7,8a-hexahydro-1H-naphthalen-1-ol
LEDER C[C@](O)(CC[C@H]1C(=C)C[C@H](O)[C@H]2C(C)(C)CCC[C@]12C)C=C
Specifikationer
Kemiskt namn eller material Larixol
Kvantitet 500 mg
InChI formel InChI=1S/C20H34O2/c1-7-19(5,22)12-9-15-14(2)13-16(21)17-18(3,4)10-8-11-20(15,17)6/h7,15-17,21-22H,1-2,8-13H2,3-6H3/t15-,16-,17-,19+,20+/m0/s1
Formel vikt 306.2559