Läs mer
Larixyl Acetate, TRC
Produktkod. 30012022
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012022 50 mg
30011196 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30012022 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL17595550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4608-49-5
Molekylformel C22 H36 O3
Molekylvikt (g/mol) 348.52
Synonym (αS,1S,4S,4aS,8aR)-4-(Acetyloxy)-α-ethenyldecahydro-α,5,5,8a-tetramethyl-2-methylene-1-naphthalenepropanol, [1S-[1α(R*),4β,4aβ,8aα]]-4-(Acetyloxy)-α-ethenyldecahydro-α,5,5,8a-tetramethyl-2-methylene-1-naphthalenepropanol
IUPAC-namn [(3S)-5-[(1S,4S,4aS,8aR)-4-hydroxy-5,5,8a-trimethyl-2-methylidene-3,4,4a,6,7,8-hexahydro-1H-naphthalen-1-yl]-3-methylpent-1-en-3-yl] acetate
LEDER CC(=O)O[C@@](C)(CC[C@H]1C(=C)C[C@H](O)[C@H]2C(C)(C)CCC[C@]12C)C=C
Specifikationer
Kemiskt namn eller material Larixyl Acetate
Kvantitet 50 mg
InChI formel InChI=1S/C22H36O3/c1-8-21(6,25-16(3)23)13-10-17-15(2)14-18(24)19-20(4,5)11-9-12-22(17,19)7/h8,17-19,24H,1-2,9-14H2,3-7H3/t17-,18-,19-,21+,22+/m0/s1
Formel vikt 348.2664