Läs mer
Laropiprant, MedChemExpress
Produktkod. 16497836
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16497836 100 mg -
16490528 5 mg -
16448068 10 mg -
16433987 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16497836 Leverantör Medchem Express Leverantörsnummer HY50175
MedChemExpress Laropiprant is a potent and selective DP receptor antagonist with Ki values of 0.57 nM and 2.95 nM for DP receptor and TP Receptor, respectively.
Laropiprant is a potent and selective DP receptor antagonist with Ki values of 0.57 nM and 2.95 nM for DP receptor and TP Receptor, respectively.
Specifikationer
Kemiskt namn eller material Laropiprant
CAS 571170-77-9
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C21H19ClFNO4S
Kvantitet 100 mg
Synonym MK-0524
Löslighetsinformation DMSO : ≥ 100 mg/mL (229.41 mM)
LEDER O=S(C1=CC(F)=CC2=C1N(C3=C2CC[C@@H]3CC(O)=O)CC4=CC=C(Cl)C=C4)(C)=O
Molekylvikt (g/mol) 435.9
Formel vikt 435.9
Procent renhet 99.73%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) COVID-19-immunoregulation