missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lass1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA112918
Additional Details : Weight : 0.01000kg
Description
A suggested positive control for this product is hypothalamus lysate.
This gene encodes a member of the bone morphogenetic protein family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site that is cleaved to produce a mature protein containing seven conserved cysteine residues. Members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. Studies in yeast suggest that the encoded protein is involved in aging. This protein is transcribed from a monocistronic mRNA as well as a bicistronic mRNA, which also encodes growth differentiation factor 1.Specifications
Lass1 | |
Polyclonal | |
PBS with 0.08% sodium azide | |
P27544 | |
CERS1 | |
Synthetic peptide corresponding to the human Lass1 protein. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
CERS1 | |
Ceramide synthase 1; CERS1; EPM8; LAG1; LAG1 homolog, ceramide synthase 1; LAG1 longevity assurance homolog 1; LASS1; longevity assurance (LAG1, S. cerevisiae) homolog 1; longevity assurance gene 1 protein homolog 1; longevity assurance homolog 1; longevity assurance-like protein 1; MGC90349; protein UOG-1; UOG1; upstream of GDF1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10715 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |