Läs mer
BRAND™ Lateral Burette Stopcock and Spare Key
Produktkod. 10375421 Handla allt Brand Produkter
Material:
Borosilikatglas (3.3)
Förpackningsstorlek:
Varje
Produktkod. 10375421
Brand: BRAND™ 81140
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Lateral Burette Stopcock and Spare Key for use with burettes with a capacity of 2 to 10 mL.
  • Stopcock housing with burette tip
  • Interchangeable
  • PTFE or borosilicate glass key with retention device
TRUSTED_SUSTAINABILITY
Specifikationer
Material Borosilikatglas (3.3)
För användning med (utrustning) 2 to 10 mL Burette
Inkluderar Securing Device