Läs mer
Lathosterol, TRC
Produktkod. 30012878
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012878 100 mg
30010990 25 mg
30011759 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30012878 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL177500100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 80-99-9
Molekylformel C27 H46 O
Molekylvikt (g/mol) 386.65
Synonym Cholest-7-en-3-ol, (3β,5α)- (9CI, ACI), (3β,5α)-Cholest-7-en-3-ol (ACI), 5α-Cholest-7-en-3β-ol (8CI), 3β-Hydroxy-5α-cholest-7-ene, 7-En-3-cholestanol, Cholest-7-en-3-ol, Lathosterol, γ-Cholestenol, δ7-Cholestenol
IUPAC-namn (3S,5S,9R,10S,13R,14R,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,5,6,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
LEDER CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2C3=CC[C@H]4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Specifikationer
Kemiskt namn eller material Lathosterol
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h10,18-21,23-25,28H,6-9,11-17H2,1-5H3/t19-,20+,21+,23-,24+,25+,26+,27-/m1/s1
Formel vikt 386.3549
Procent renhet >95%