Läs mer
Lauroyl-L-carnitine Hydrochloride, TRC
Produktkod. 30483208
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30483208 1 g
30474669 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30483208 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL1786801G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6919-91-1
Molekylformel C19H38ClNO4
Molekylvikt (g/mol) 379.96
Synonym (2R)-3-Carboxy-N,N,N-trimethyl-2-[(1-oxododecyl)oxy]-1-propanaminium Chloride, (R)-3-Carboxy-N,N,N-trimethyl-2-[(1-oxododecyl)oxy]-1-propanaminium Chloride, (-)-(3-Carboxy-2-hydroxypropyl)trimethyl-ammonium Chloride Laurate, (-)-Lauric Acid Ester with (3-Carboxy-2-hydroxypropyl)trimethylammonium Chloride
IUPAC-namn [(2R)-3-carboxy-2-dodecanoyloxypropyl]-trimethylazanium;chloride
LEDER O=C(CCCCCCCCCCC)O[C@H](CC(O)=O)C[N+](C)(C)C.[Cl-]
Specifikationer
Kemiskt namn eller material Lauroyl-L-carnitine Hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C19H37NO4.ClH/c1-5-6-7-8-9-10-11-12-13-14-19(23)24-17(15-18(21)22)16-20(2,3)4;/h17H,5-16H2,1-4H3;1H/t17-;/m1./s1
Formel vikt 379.2489
Procent renhet >95%