missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lauroylamide Propylbetaine, TRC
Specifications
Specifications
| Kemiskt namn eller material | Lauroylamide Propylbetaine |
| CAS | 4292-10-8 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C19H38N2O3 |
| Kvantitet | 250 mg |
| Synonym | N-(Carboxymethyl)-N,N-dimethyl-3-[(1-oxododecyl)amino]-1-propanaminium Inner Salt, (Carboxymethyl)(3-lauramidopropyl)dimethylammonium Hydroxide Inner Salt, N-(Carboxymethyl)-N,N-dimethyl-3-[(1-oxododecyl)amino]-1-propanaminium Hydroxide Inner Salt, (3-Lauramidopropyl)dimethylbetaine, Amphitol 20AB, Amphosol LB, Amphoteric L, Anpholex LB 2, Dehyton L 12, Dimethyl(lauramidopropyl)betaine, N,N-Dimethyl-N-dodecanoylaminopropylbetaine, Obazoline CAB, Obazoline LAB, Rewoteric AMB 12, Rikabion B 300, Softazoline LPB, Softazoline LPB-R, Tego-Betain L 90 |
| InChI formel | InChI=1S/C19H38N2O3/c1-4-5-6-7-8-9-10-11-12-14-18(22)20-15-13-16-21(2,3)17-19(23)24/h4-17H2,1-3H3,(H-,20,22,23,24) |
| LEDER | CCCCCCCCCCCC(=O)NCCC[N+](C)(C)CC(=O)[O-] |
| IUPAC-namn | 2-((3-dodecanamidopropyl)dimethylammonio)acetate |
| Molekylvikt (g/mol) | 342.52 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?