missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LBP Recombinant Human Protein, His Tag
LBP Recombinant Human Protein, His Tag
Brand: Invitrogen 10526H08H25
Additional Details : Weight : 0.01000kg
Description
Lipopolysaccharide Binding Protein (LBP), recombinant human protein is supplied in a frozen format. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.This recombinant protein was expressed from a DNA sequence encoding the human LBP (NP_004130.2) extracellular domain (Met 1-Val 481) fused to the a polyhistidine tag at the C-terminus.
Activity: The activity of this protein has not been tested.
Formulation: Lyophilized in 20 mM Tris, 100 mM NaCl, 0.05 mM EDTA, 10% glycerol, pH 8.0.
Reconstitution: No reconstitution is needed. Store aliquots under sterile conditions at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles.
This protein is manufactured by Sino Biological Inc.
Specifications
Protein Assays and Analysis, Assay Standard | |
3929 | |
LBP Recombinant Human Protein | |
5 x 5μg | |
For research use only. Not for use in diagnostic procedures | |
MGC22233, LBP | |
5 Vials of protein in suspension buffer | |
LBP Recombinant Human Protein | |
His Tag | |
Other proteins | |
Other proteins |
Frozen | |
52.5kDa | |
PAGE | |
–80°C | |
5 Pack(s) | |
LBP Recombinant Human Protein | |
LBP | |
Human | |
Mammalian | |
Frozen | |
≥95% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.