missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LCA5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19333-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The MW of this protein is 80 kDa, and this antibody specifically recognizes the 80 kDa protein.
Might be involved in minus end-directed microtubule transport.Specifications
LCA5 | |
Polyclonal | |
Unconjugated | |
LCA5 | |
C6orf152, LCA5, Leber congenital amaurosis 5, Lebercilin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
167691 | |
-20°C | |
Liquid |
Western Blot | |
0.42 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q86VQ0 | |
LCA5 | |
LCA5 Fusion Protein Ag5140 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |