missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LCA5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | LCA5 |
---|---|
Concentration | 0.42 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16878954
|
Proteintech
19333-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16868954
|
Proteintech
19333-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The MW of this protein is 80 kDa, and this antibody specifically recognizes the 80 kDa protein.
Might be involved in minus end-directed microtubule transport.Specifications
LCA5 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q86VQ0 | |
167691 | |
LCA5 Fusion Protein Ag5140 | |
Primary | |
-20°C | |
LCA5 |
0.42 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C6orf152, LCA5, Leber congenital amaurosis 5, Lebercilin | |
LCA5 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |