Läs mer
LCL521, MedChemExpress
Produktkod. 16474896
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16474896 100 mg -
16470237 50 mg 50mg
16443868 10 mg -
16416597 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16474896 Leverantör Medchem Express Leverantörsnummer HY103593
MedChemExpress LCL521 is an acid ceramidase (ACDase) inhibitor. LCL521 also inhibits the lysosomal acid sphingomyelinase (ASMase).
LCL521 is an acid ceramidase (ACDase) inhibitor. LCL521 also inhibits the lysosomal acid sphingomyelinase (ASMase).
Specifikationer
Kemiskt namn eller material LCL521
Fysisk form Oil
CAS 1226851-11-1
Färg Light Yellow
Kvantitet 100 mg
Anteckningar om renhetsgrad Research
Molekylformel C31H52N4O7
För användning med (applikation) Cancer-programmed cell death
Löslighetsinformation DMSO : 165 mg/mL (278.35 mM; Need ultrasonic) ∣Ethanol : 100 mg/mL (168.70 mM; Need ultrasonic)
LEDER O=C(CN(C)C)O[C@H](C1=CC=C([N+]([O-])=O)C=C1)[C@@H](COC(CN(C)C)=O)NC(CCCCCCCCCCCCC)=O
Molekylvikt (g/mol) 592.77
Formel vikt 592.77