Läs mer
LCZ696, TRC
Produktkod. 18080620
Change view
Kvantitet:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18080620 5 mg
18071620 50 mg
18010830 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 18080620 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL2700055MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1884447-69-1
Molekylformel C24 H28 N O5 . C24 H27 N5 O3 . 3Na . x(H2O)
Molekylvikt (g/mol) 327.4
Synonym LCZ696, N-(1-Oxopentyl)-N-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-L-valine, compound with a-Ethyl (aR,gS)-g-[(3-carboxy-1-oxopropyl)amino]-a-methyl[1,1'-biphenyl]-4-pentanoate, Sodium Salt, Hydrate (2:2:6:5), Entresto, Sacubitril-valsartan
IUPAC-namn trisodium;4-[[(1S,3R)-4-ethoxy-3-methyl-4-oxo-1-[(4-phenylphenyl)methyl]butyl]amino]-4-oxo-butanoate;(2S)-3-methyl-2-[pentanoyl-[[4-[2-(1,2,3-triaza-4-azanidacyclopenta-2,5-dien-5-yl)phenyl]phenyl]methyl]amino]butanoate;hydrate
LEDER O.[Na+].[Na+].[Na+].CCCCC(=O)N(Cc1ccc(cc1)c2ccccc2c3nnn[n-]3)[C@@H](C(C)C)C(=O)[O-].CCOC(=O)[C@H](C)C[C@@H](Cc4ccc(cc4)c5ccccc5)NC(=O)CCC(=O)[O-]
Specifikationer
Kemiskt namn eller material Sacubitril-valsartan Mixture
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C24H29N5O3.C24H29NO5.3Na.H2O/c1-4-5-10-21(30)29(22(16(2)3)24(31)32)15-17-11-13-18(14-12-17)19-8-6-7-9-20(19)23-25-27-28-26-23;1-3-30-24(29)17(2)15-21(25-22(26)13-14-23(27)28)16-18-9-11-20(12-10-18)19-7-5-4-6-8-19;;;;/h6-9,11-14,16,22H,4-5,10,15H2,1-3H3,(H2,25,26,27,28,31,32);4-12,17,21H,3,13-16H2,1-2H3,(H,25,26)(H,27,28);;;;1H2/q;;3*+1;/p-3/t22-;17-,21+;;;;/m01..../s1
Procent renhet >95%