Läs mer
Tocris Bioscience™ LDC 000067
Kvantitet:
5 mg
25 mg
Beskrivning
LDC 000067 is a potent and selective CDK9 inhibitor (IC50 = 44 nM at CDK9-cyclin T1). Inhibits other CDKs in the micromolar range (IC50 values are 2.4, 5.5, 9.2, >10 and >10 μM for CDK2-cyclin A, CDK1-cyclin B1, CDK4-cyclin D1, CDK6-cyclin D3 and CDK7-cyclin H-MAT1, respectively). Inhibits P-TEFb-dependent in vitro transcription. Induces apoptosis in vitro and in vivo in combination with BI 894999.
Specifikationer
Specifikationer
| Kemiskt namn eller material | 3-[[6-(2-Methoxyphenyl)-4-pyrimidinyl]amino]benzenemethanesulfonamide |
| CAS | 1073485-20-7 |
| Kvantitet | 25 mg |
| Molekylformel | C18H18N4O3S |
| Formel vikt | 370.43 |
| Renhet | 98% |