missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LDHA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19987-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The antibody has no cross reaction to LDHB and LDHC. This antibody is specific to LDHA.
Lactate dehydrogenase (LDH) is the enzyme in the glycolytic pathway that converts pyruvate to lactate with concomitant interconversion of NADH and NAD+. In mammals, the enzyme is encoded by three genes: LDHA (M or muscle form), LDHB (H or heart form), and LDHC (X or testis form). The three human LDHs have 84-89% sequence similarities and 69-75% amino acid identities. There are five different LDH isoenzymes based on the proportion of M and H chains existing in the LDH tetrameric structure. The LDHA is hypoxia inducible and its expression is directly controlled by the transcriptional activity of the hypoxia inducible factor 1a (HIF1a). LDHB is also known to be upregulated in many cancers.Specifications
LDHA | |
Polyclonal | |
Unconjugated | |
LDHA | |
lactate dehydrogenase A, LDH A, LDH M, LDH muscle subunit, LDH1, LDHA, PIG19 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16828, 24533, 3939 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P00338, P04642, P06151 | |
LDHA | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |