missing translation for 'onlineSavingsMsg'
Learn More

LDLRAD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 20096-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16869574

  • 4721.67 SEK / 150µL
Estimated Shipment: 21-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The antibody can recognize all 3 isoforms of LDLRAD1.

LDLRAD1 belongs to the LDLR family.
Specifications

Specifications

LDLRAD1
Polyclonal
Unconjugated
LDLRAD1
LDLRAD1
Rabbit
Antigen Affinity Chromatography
RUO
388633, 546840
-20°C
Liquid
Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5T700
LDLRAD1
Peptide
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers