Läs mer
LDN 193189 Hydrochloride, TRC
Produktkod. 30415615
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30415615 10 mg
30413769 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30415615 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP48013310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material LDN 193189 Hydrochloride
CAS 2648112-58-5
Analyt- eller komponentnamn LDN 193189 Hydrochloride
Frakt skick Room Temperature
Molekylformel C25H22N6 • x(HCl)
Synonym 4-(6-(4-(Piperazin-1-yl)phenyl)pyrazolo[1,5-a]pyrimidin-3-yl)quinoline Hydrochoride
InChI formel InChI=1S/C25H22N6/c1-2-4-24-22(3-1)21(9-10-27-24)23-16-29-31-17-19(15-28-25(23)31)18-5-7-20(8-6-18)30-13-11-26-12-14-30/h1-10,15-17,26H,11-14H2
Lösningstyp Neat
LEDER O=C([C@@H](N)CC1=CC(S(C)(=O)=O)=CC=C1)OCC2=CC=CC=C2.Cl
Industry Type Pharmaceutical
Molekylvikt (g/mol) 406.483646