missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
STEMCELL Technologies LDN193189 (Dihydrochloride)
Beskrivning
- LDN193189 is a potent inhibitor of the bone morphogenetic (BMP) pathway, inhibiting ALK1, ALK2, ALK3, and ALK6 (IC50 = 0.8, 0.8, 5.3, and 16.7 nM respectively; Sanvitale et al.) It is a derivative of Dorsomorphin (Catalog #72102) that is typically used at approximately 100-fold lower concentrations (Sanvitale et al.; Vogt et al.) This product is supplied as the dihydrochloride salt of the molecule and it is the most soluble version of LDN193189
- Promotes differentiation of neural progenitor cells from human pluripotent stem cells (Chambers et al.; Kriks et al.)
- Promotes differentiation of neural crest cells from human pluripotent stem cells (Kreitzer et al.)
- Promotes differentiation of anterior foregut endoderm from human and mouse pluripotent stem cell-derived definitive endoderm (Kearns et al.)
- Promotes differentiation of sensory epithelial cells of the inner ear from mouse embryonic stem cells (Koehler et al.)
- Inhibits prostate and breast cancer tumor growth (Balboni et al.; Lee et al.)
- Prevents ovarian cancer cell proliferation (Tsai et al.)
Specifikationer
Specifikationer
| CAS | 1435934-00-1 |
| Synonym | DM-3189 |
| Kvantitet | 10 mg |
| Molekylformel | C25H22N6 · 2HCl |
| Renhet | >98% |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?