Läs mer
LDN193189 Tetrahydrochloride, MedChemExpress
Produktkod. 16491217
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16491217 50 mg
16467707 5 mg
16424978 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16491217 Leverantör Medchem Express Leverantörsnummer HY12071A
MedChemExpress LDN193189 Tetrahydrochloride is a selective BMP type I receptor inhibitor, which efficiently inhibits ALK2 and ALK3 (IC50=5 nM and 30 nM, respectively), with weaker effects on ALK4, ALK5 and ALK7 (IC50≥500 nM).
LDN193189 Tetrahydrochloride is a selective BMP type I receptor inhibitor, which efficiently inhibits ALK2 and ALK3 (IC50=5 nM and 30 nM, respectively), with weaker effects on ALK4, ALK5 and ALK7 (IC50≥500 nM).
Specifikationer
Kemiskt namn eller material LDN193189 Tetrahydrochloride
CAS 2310134-98-4
Färg Off-Yellow
Anteckningar om renhetsgrad Research
Molekylformel C25H26Cl4N6
Kvantitet 50 mg
Löslighetsinformation H2O : 33.33 mg/mL (60.34 mM; Need ultrasonic) ∣DMSO : 4.17 mg/mL (7.55 mM; Need ultrasonic)
LEDER C1(C2=C3N=CC(C4=CC=C(N5CCNCC5)C=C4)=CN3N=C2)=CC=NC6=CC=CC=C16.Cl.Cl.Cl.Cl
Molekylvikt (g/mol) 552.33
Formel vikt 552.33
Procent renhet 98.04%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-Kinase/protease