Learn More
BRAND™ LDPE Bottle Closure
Description
- Includes dropper nozzle
- For use with threaded bottles
Specifications
Closure Size | GL 14 |
For Use With (Equipment) | Threaded Bottles |
Includes | Dropper Nozzle |
Material | LDPE |