Learn More
Kautex™ 250 mL Safety Wash Bottles Series 303 LDPE
Designed to give an accurate fine controlled stream of liquid. Kautex™ 250mL Safety Wash Bottles provide excellent chemical resistance for laboratory safety.
Brand: Kautex™ 2000770017
Additional Details : Weight : 0.03600kg
Description
- Imprinted with hazard symbols and substances of content in German, English and French language
- Writing space
- Abrasion-resistant printing
- Closure color matches color of ink for easy identification of media
- Leak-proof
- Completely emptied by sliding delivery tube, even when inverted.
Specifications
Wash Bottle | |
64 mm | |
Isopropanol | |
Natural | |
LDPE | |
150 Pk. | |
Wide |
120 mm | |
Screw Cap with Delivery Nozzle | |
250 mL | |
30.8 mm | |
10 | |
Yellow | |
Round |