missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brand™ LDPE Wash Bottles with Venting Valve for Acetone
Click to view available options
Capacity (Metric):
250 mL
Description
- For distilled water and organic solvents only
- Large opening for convenient filling
- Label with designation of contents and general safety statements in English (was CAS numbers and NFPA codes) and German (was CAS numbers)
- Manufactured from flexible PE-LD, limited to temperatures below 80°C
Specifications
Capacity (Metric) | 250 mL |
Chemical Label | Acetone |
Material | LDPE |
Type | Wash Bottle |
Closure Color | Red |
Closure Included | Yes |
Closure Type | Stopper |
Mouth | Wide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'