Läs mer
Brand™ LDPE Wash Bottles without Venting Valve
Handla allt Brand Produkter
missing translation for 'orderingAttributeHoverText'
Kemisk etikett:
Isopropanol
Methanol
missing translation for 'unitSize'
Paket med 5
Description
- For distilled water and organic solvents only
- Large opening for convenient filling
- Label with designation of contents and general safety statements in English (was CAS numbers and NFPA codes) and German (was CAS numbers)
- Manufactured from flexible PE-LD, limited to temperatures below 80°C
Specifications
Specifications
| Kapacitet (metrisk) | 250 mL |
| Material | LDPE |
| Stängningsfärg | Blue |
| Mun | Wide |
| Produkttyp | Wash Bottle |
| Kemisk etikett | Isopropanol |